α-Bromo-2-chlorotoulene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide

001008-25G 001008-100G
FLUO001008-25GEA 0 GBP
FLUO001008-25G FLUO001008-100G
α-Bromo-2-chlorotoulene
α-Bromo-2-chlorotoulene
Formula: C₇H₆BrCl
Boiling Pt: 235 °C (1013 hPa)
Density: 1.5897 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000566
CAS Number: 611-17-6
EINECS: 210-257-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR