4-tert-Butyl-1-bromobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001068.100G 001068-250G 001068-50G
FLUO001068.100GEA 0 GBP
FLUO001068.100G FLUO001068-250G FLUO001068-50G
4-tert-Butyl-1-bromobenzene
4-tert-Butyl-1-bromobenzene
Formula: C₁₀H₁₃Br
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 15.5 °C
Density: 1.2572 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000108
CAS Number: 3972-65-4
EINECS: 223-599-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR