α,3-Dibromotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene

001367-50G 001367-250G
FLUO001367-50GEA 0 GBP
FLUO001367-50G FLUO001367-250G
α,3-Dibromotoluene
α,3-Dibromotoluene
Formula: C₇H₆Br₂
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 41…44 °C
Density: 1.56 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000176
CAS Number: 823-78-9
EINECS: 212-519-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR