1,4,8,11-Tetraazacyclotetradecane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: CYCLAM

259160-1G 259160-5G
SIAL259160-1GEA 64.6 GBP
SIAL259160-1G SIAL259160-5G
1,4,8,11-Tetraazacyclotetradecane, Sigma-Aldrich®
1,4,8,11-Tetraazacyclotetradecane
Formula: C₁₀H₂₄N₄
MW: 200.33 g/mol
MDL Number: MFCD00005105
CAS Number: 295-37-4
EINECS: 206-039-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR