1-Benzyloxy-3-bromobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Benzyloxybromobenzene, 1-(Benzyloxy)-3-bromobenzene

023255-25G 023255-5G
FLUO023255-25GEA 0 GBP
FLUO023255-25G FLUO023255-5G
1-Benzyloxy-3-bromobenzene
1-Benzyloxy-3-bromobenzene
Formula: C₁₃H₁₁BrO
MW: 263.13
MDL Number: MFCD00155065
CAS Number: 53087-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR