2,6-Dimethoxy-3-pyridineboronic acid pinacol ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021611-5G 021611-1G
FLUO021611-5GEA 0 GBP
FLUO021611-5G FLUO021611-1G
2,6-Dimethoxy-3-pyridineboronic acid pinacol ester
2,6-Dimethoxy-3-pyridineboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₄
MDL Number: MFCD03788240
CAS Number: 214360-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR