1-Benzyloxy-4-chlorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
018221-10G
018221-25G
FLUO018221-10GEA
0
GBP
FLUO018221-10G
FLUO018221-25G
1-Benzyloxy-4-chlorobenzene
1-Benzyloxy-4-chlorobenzene
Formula:
C₁₃H₁₁ClO |
MDL Number:
MFCD00079702 CAS Number: 7700-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...