1-Benzyloxy-4-bromobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018220-1G 018220-10G 018220-100G 018220-25G
FLUO018220-1GEA 0 GBP
FLUO018220-1G FLUO018220-10G FLUO018220-100G FLUO018220-25G
1-Benzyloxy-4-bromobenzene
1-Benzyloxy-4-bromobenzene
Formula: C₁₃H₁₁BrO
MDL Number: MFCD00028016
CAS Number: 6793-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR