(2-Nitro-4-(trifluoromethyl)phenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

010345-1G 010345-5G
FLUO010345-1GEA 0 GBP
FLUO010345-1G FLUO010345-5G
(2-Nitro-4-(trifluoromethyl)phenyl)acetic acid
(2-Nitro-4-(trifluoromethyl)phenyl)acetic acid
Formula: C₉H₆F₃NO₄
MDL Number: MFCD00091534
CAS Number: 1735-91-7
EINECS: 217-081-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR