1-Benzyloxy-4-fluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Benzyloxy)-4-fluorobenzene

009022-25G 009022-5G
FLUO009022-25GEA 0 GBP
FLUO009022-25G FLUO009022-5G
1-Benzyloxy-4-fluorobenzene
1-Benzyloxy-4-fluorobenzene
Formula: C₁₃H₁₁FO
MW: 202.23
MDL Number: MFCD03788273
CAS Number: 370-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR