(2-Chloro-3,6-difluorophenyl)acetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005606-1G 005606-5G
FLUO005606-1GEA 0 GBP
FLUO005606-1G FLUO005606-5G
(2-Chloro-3,6-difluorophenyl)acetic acid
(2-Chloro-3,6-difluorophenyl)acetic acid
Formula: C₈H₅ClF₂O₂
MW: 206.58 g/mol
MDL Number: MFCD01631448
CAS Number: 261762-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR