2-(Cyclopentyloxy)-3-pyridineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Cyclopentyloxypyridine-3-boronic acid pinacol ester

PN-8895-1G PN-8895-250M
COBBPN-8895-1GEA 0 GBP
COBBPN-8895-1G COBBPN-8895-250M
2-(Cyclopentyloxy)-3-pyridineboronic acid pinacol ester
2-(Cyclopentyloxy)-3-pyridineboronic acid pinacol ester
Formula: C₁₆H₂₄BNO₃
MDL Number: MFCD07781170
CAS Number: 1073371-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR