2-Isopropoxy-5-pyridineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(6-Isopropoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Isopropoxy-3-pyridylboronic acid Pinacol ester, 2-Isopropoxypyridine-5-boronic acid pinacol ester

PN-8890-1G PN-8890-5G
COBBPN-8890-1GEA 0 GBP
COBBPN-8890-1G COBBPN-8890-5G
2-Isopropoxy-5-pyridineboronic acid pinacol ester
2-Isopropoxy-5-pyridineboronic acid pinacol ester
Formula: C₁₄H₂₂BNO₃
MW: 263.14 g/mol
MDL Number: MFCD07368871
CAS Number: 871839-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR