2-Phenylpropylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Phenyl-1-propylboronic acid pinacol ester, 4,4,5,5-Tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane

PN-8828-5G PN-8828-1G
COBBPN-8828-5GEA 0 GBP
COBBPN-8828-5G COBBPN-8828-1G
2-Phenylpropylboronic acid pinacol ester
2-Phenylpropylboronic acid pinacol ester
Formula: C₁₅H₂₃BO₂
MW: 246.16 g/mol
MDL Number: MFCD09953475
CAS Number: 280559-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR