2,6-Difluoro-3,5-dipyridineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,6-Difluoro-3,5-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-6018-1G PN-6018-250M
COBBPN-6018-1GEA 0 GBP
COBBPN-6018-1G COBBPN-6018-250M
2,6-Difluoro-3,5-dipyridineboronic acid pinacol ester
2,6-Difluoro-3,5-dipyridineboronic acid pinacol ester
Formula: C₁₇H₂₅B₂F₂NO₄
MW: 367.01 g/mol
MDL Number: MFCD12913982
CAS Number: 1218789-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR