2-Acetamido-5-pyridineboronic acid pinacol ester hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)acetamide hydrochloride

PN-5727-1G PN-5727-250M
COBBPN-5727-1GEA 0 GBP
COBBPN-5727-1G COBBPN-5727-250M
2-Acetamido-5-pyridineboronic acid pinacol ester hydrochloride
2-Acetamido-5-pyridineboronic acid pinacol ester hydrochloride
Formula: C₁₃H₂₀BClN₂O₃
MW: 298.58 g/mol
MDL Number: MFCD12026097
CAS Number: 1150561-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR