3-Fluoro-4-pyridineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Fluoropyridine-4-boronic acid pinacol ester

PN-3440-5G PN-3440-1G
COBBPN-3440-5GEA 0 GBP
COBBPN-3440-5G COBBPN-3440-1G
3-Fluoro-4-pyridineboronic acid pinacol ester
3-Fluoro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
MDL Number: MFCD06798251
CAS Number: 458532-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR