5-Bromo-3-pyridineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Bromopyridine-5-boronic acid pinacol ester

PN-3426-5G PN-3426-1G
COBBPN-3426-5GEA 0 GBP
COBBPN-3426-5G COBBPN-3426-1G
5-Bromo-3-pyridineboronic acid pinacol ester
5-Bromo-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BBrNO₂
MW: 283.96 g/mol
MDL Number: MFCD06798248
CAS Number: 452972-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR