2-Fluoro-4-pyridineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoropyridine-4-boronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-2916-5G PN-2916-1G
COBBPN-2916-5GEA 0 GBP
COBBPN-2916-5G COBBPN-2916-1G
2-Fluoro-4-pyridineboronic acid pinacol ester
2-Fluoro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
MDL Number: MFCD06798253
CAS Number: 458532-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR