1-Benzyloxy-4-bromobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Benzyloxybromobenzene, 1-Bromo-4-(phenylmethoxy)benzene

OT-0693-25G OT-0693-100G OT-0693-500G
COBBOT-0693-25GEA 0 GBP
COBBOT-0693-25G COBBOT-0693-100G COBBOT-0693-500G
1-Benzyloxy-4-bromobenzene
1-Benzyloxy-4-bromobenzene
Formula: C₁₃H₁₁BrO
MW: 263.13
MDL Number: MFCD00028016
CAS Number: 6793-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR