2-Bromo-6-chlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Bromo-6-chlorophenyl)boronic acid

FA-1571-1G FA-1571-25G
COBBFA-1571-1GEA 0 GBP
COBBFA-1571-1G COBBFA-1571-25G
2-Bromo-6-chlorophenylboronic acid
2-Bromo-6-chlorophenylboronic acid
Formula: C₆H₅BBrClO₂
MW: 235.27 g/mol
MDL Number: MFCD08701777
CAS Number: 1107580-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR