2-Bromo-5-chlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Bromo-5-chlorophenyl)boronic acid

BB-6002-1G BB-6002-5G BB-6002-250M
COBBBB-6002-1GEA 0 GBP
COBBBB-6002-1G COBBBB-6002-5G COBBBB-6002-250M
2-Bromo-5-chlorophenylboronic acid
2-Bromo-5-chlorophenylboronic acid
Formula: C₆H₅BBrClO₂
MW: 235.27 g/mol
MDL Number: MFCD12026715
CAS Number: 1217501-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR