2,6-Difluoro-4-hydroxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2,6-Difluoro-4-hydroxyphenyl)boronic acid

BB-5094-5G BB-5094-1G
COBBBB-5094-5GEA 0 GBP
COBBBB-5094-5G COBBBB-5094-1G
2,6-Difluoro-4-hydroxyphenylboronic acid
2,6-Difluoro-4-hydroxyphenylboronic acid
Formula: C₆H₅BF₂O₃
MW: 173.91 g/mol
MDL Number: MFCD09878344
CAS Number: 957065-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR