2-Fluoro-3-methoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-3-methoxybenzeneboronic acid, (2-Fluoro-3-methoxyphenyl)boronic acid

BB-2460-5G BB-2460-1G
COBBBB-2460-5GEA 0 GBP
COBBBB-2460-5G COBBBB-2460-1G
2-Fluoro-3-methoxyphenylboronic acid
2-Fluoro-3-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD02094728
CAS Number: 352303-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR