2-Chloro-3-fluorophenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Chloro-3-fluorobenzeneboronic acid, (2-Chloro-3-fluorophenyl)boronic acid
BB-2254-5G
BB-2254-1G
COBBBB-2254-5GEA
0
GBP
COBBBB-2254-5G
COBBBB-2254-1G
2-Chloro-3-fluorophenylboronic acid
2-Chloro-3-fluorophenylboronic acid
Formula:
C₆H₅BClFO₂ MW: 174.37 g/mol |
MDL Number:
MFCD07363741 CAS Number: 871329-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...