2-Chloro-5-fluorophenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Chloro-5-fluorobenzeneboronic acid
BB-2251-5G
BB-2251-25G
BB-2251-1G
COBBBB-2251-5GEA
0
GBP
COBBBB-2251-5G
COBBBB-2251-25G
COBBBB-2251-1G
2-Chloro-5-fluorophenylboronic acid
2-Chloro-5-fluorophenylboronic acid
Formula:
C₆H₅BClFO₂ |
MDL Number:
MFCD06656272 CAS Number: 444666-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...