4-Nitrophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane, 4-Nitrophenylboronic acid Pinacol ester, 1-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 4-Nitrobenzeneboronic acid pinacol ester

BB-2106-25G BB-2106-1G BB-2106-5G
COBBBB-2106-25GEA 0 GBP
COBBBB-2106-25G COBBBB-2106-1G COBBBB-2106-5G
4-Nitrophenylboronic acid pinacol ester
4-Nitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BNO₄
MDL Number: MFCD02179437
CAS Number: 171364-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR