Allylpentafluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-(Pentafluorophenyl)-1-propene, 1-Allyl-2,3,4,5,6-pentafluorobenzene

003947-25G 003947-5G
FLUO003947-25GEA 0 GBP
FLUO003947-25G FLUO003947-5G
Allylpentafluorobenzene
Allylpentafluorobenzene
Formula: (C₆F₅)CH₂CHCH₂
Boiling Pt: 148…149 °C (1013 hPa)
Density: 1.35 g/cm³ (20 °C)
MDL Number: MFCD00000301
CAS Number: 1736-60-3
EINECS: 217-083-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR