Benzene, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

12540-50ML-F 12540-5ML-F
12540-50ML-FEA 322 GBP
12540-50ML-F 12540-5ML-F
Benzene, Supelco®
Benzene
Formula: C₆H₆
Boiling Pt: 80.1 °C (1013 hPa)
Melting Pt: 5.5 °C
Density: 0.878 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00003009
CAS Number: 71-43-2
EINECS: 200-753-7
UN: 1114
ADR: 3,II
REACH: 01-2119447106-44
Merck Index: 13,01066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=195f967e9e53065b2ed6eadc5ca12dfa3f7930ed&vid=b973b9785fd03a80a3bef7ce0efb171436ea53eb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR