4'-Methylbiphenyl-4-carboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4'-Methyl-[1,1'-biphenyl]-4-carboxylic acid, 4'-Methyl-4-biphenylcarboxylic acid

OT-8003-5G OT-8003-1G
COBBOT-8003-5GEA 0 GBP
COBBOT-8003-5G COBBOT-8003-1G
4'-Methylbiphenyl-4-carboxylic acid
4'-Methylbiphenyl-4-carboxylic acid
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD00448778
CAS Number: 720-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR