2',5'-Dimethylbiphenyl-3-carboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4898-1G
CA-4898-5G
COBBCA-4898-1GEA
0
GBP
COBBCA-4898-1G
COBBCA-4898-5G
2',5'-Dimethylbiphenyl-3-carboxylic acid
2',5'-Dimethylbiphenyl-3-carboxylic acid
Formula:
C₁₅H₁₄O₂ |
MDL Number:
MFCD11938175 CAS Number: 1181626-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...