5-Fluoro-2-iodobenzotrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024520-25G 024520-5G
FLUO024520-25GEA 0 GBP
FLUO024520-25G FLUO024520-5G
5-Fluoro-2-iodobenzotrifluoride
5-Fluoro-2-iodobenzotrifluoride
Formula: C₇H₃F₄I
MDL Number: MFCD07369008
CAS Number: 41860-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR