4-(2-(Dimethylamino)ethyl)phenol hydrochloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008895-25G
008895-100G
FLUO008895-25GEA
0
GBP
FLUO008895-25G
FLUO008895-100G
4-(2-(Dimethylamino)ethyl)phenol hydrochloride
4-(2-(Dimethylamino)ethyl)phenol hydrochloride
Formula:
C₁₀H₁₆ClNO MW: 201.7 g/mol |
MDL Number:
MFCD00040807 CAS Number: 6027-23-2 EINECS: 227-892-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...