2-Amino-5-methylphenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-5-methylphenyboronic acid, pinacol ester, 4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

PN-8880-1G PN-8880-250M
COBBPN-8880-1GEA 0 GBP
COBBPN-8880-1G COBBPN-8880-250M
2-Amino-5-methylphenylboronic acid pinacol ester
2-Amino-5-methylphenylboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₂
MDL Number: MFCD06795664
CAS Number: 948592-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR