4-Amino-2-fluorophenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-8921-1G PN-8921-5G
COBBPN-8921-1GEA 0 GBP
COBBPN-8921-1G COBBPN-8921-5G
4-Amino-2-fluorophenylboronic acid pinacol ester
4-Amino-2-fluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₇BFNO₂
MDL Number: MFCD09951877
CAS Number: 819057-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR