2-Bromo-4-(trifluoromethoxy)benzaldehyde
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4793-1G
CA-4793-5G
COBBCA-4793-1GEA
0
GBP
COBBCA-4793-1G
COBBCA-4793-5G
2-Bromo-4-(trifluoromethoxy)benzaldehyde
2-Bromo-4-(trifluoromethoxy)benzaldehyde
Formula:
C₈H₄BrF₃O₂ |
MDL Number:
MFCD09835093 CAS Number: 1114808-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...