2-(3-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-2325-1G PN-2325-5G
COBBPN-2325-1GEA 0 GBP
COBBPN-2325-1G COBBPN-2325-5G
2-(3-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(3-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₉BO₃
MDL Number: MFCD05863915
CAS Number: 325142-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR