Isopropylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3899-1G BB-3899-5G BB-3899-25G
COBBBB-3899-1GEA 0 GBP
COBBBB-3899-1G COBBBB-3899-5G COBBBB-3899-25G
Isopropylboronic acid
Isopropylboronic acid
Formula: C₃H₉BO₂
MW: 87.91 g/mol
MDL Number: MFCD01319021
CAS Number: 80041-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR