Isopropylboronic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
648787-10G
648787-1G
SIAL648787-10GEA
436
GBP
SIAL648787-10G
SIAL648787-1G
Isopropylboronic acid, Sigma-Aldrich®
Isopropylboronic acid
Formula:
C₃H₉BO₂ MW: 87.91 g/mol |
MDL Number:
MFCD01319021 CAS Number: 80041-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...