1-Propylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Propylboronic acid

BB-8313-1G BB-8313-5G
COBBBB-8313-1GEA 0 GBP
COBBBB-8313-1G COBBBB-8313-5G
1-Propylboronic acid
1-Propylboronic acid
Formula: C₃H₉BO₂
MW: 87.91 g/mol
MDL Number: MFCD01074564
CAS Number: 17745-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR