1-Butylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2143-5G BB-2143-25G BB-2143-100G
COBBBB-2143-5GEA 0 GBP
COBBBB-2143-5G COBBBB-2143-25G COBBBB-2143-100G
1-Butylboronic acid
1-Butylboronic acid
Formula: C₄H₁₁BO₂
MDL Number: MFCD00002106
CAS Number: 4426-47-5
EINECS: 224-607-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR