1-Butylboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010773-5G
010773-25G
FLUO010773-5GEA
0
GBP
FLUO010773-5G
FLUO010773-25G
1-Butylboronic acid
1-Butylboronic acid
Formula:
C₄H₁₁BO₂ |
MDL Number:
MFCD00002106 CAS Number: 4426-47-5 EINECS: 224-607-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...