4'-Fluorobiphenyl-4-carboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4'-Fluoro-[1,1'-biphenyl]-4-carboxylic acid, 4-(4-Fluorophenyl)benzoic acid

014262-100MG 014262-1G
FLUO014262-100MGEA 0 GBP
FLUO014262-100MG FLUO014262-1G
4'-Fluorobiphenyl-4-carboxylic acid
4'-Fluorobiphenyl-4-carboxylic acid
Formula: C₁₃H₉FO₂
MW: 216.21 g/mol
MDL Number: MFCD01631909
CAS Number: 5731-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR