4'-Fluorobiphenyl-3-carboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4823-5G CA-4823-1G
COBBCA-4823-5GEA 0 GBP
COBBCA-4823-5G COBBCA-4823-1G
4'-Fluorobiphenyl-3-carboxylic acid
4'-Fluorobiphenyl-3-carboxylic acid
Formula: C₁₃H₉FO₂
MW: 216.21 g/mol
MDL Number: MFCD00452724
CAS Number: 10540-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR