3'-Fluorobiphenyl-2-carboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011490-500MG 011490-1G
FLUO011490-500MGEA 0 GBP
FLUO011490-500MG FLUO011490-1G
3'-Fluorobiphenyl-2-carboxylic acid
3'-Fluorobiphenyl-2-carboxylic acid
Formula: C₁₃H₉FO₂
MW: 216.21 g/mol
MDL Number: MFCD03426488
CAS Number: 2094-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR