3'-Methoxybiphenyl-3-carboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3'-Methoxy-[1,1'-biphenyl]-3-carboxylic acid

CA-4820-1G CA-4820-5G
COBBCA-4820-1GEA 0 GBP
COBBCA-4820-1G COBBCA-4820-5G
3'-Methoxybiphenyl-3-carboxylic acid
3'-Methoxybiphenyl-3-carboxylic acid
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD03424598
CAS Number: 168618-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR