5'-Methoxy-2'-methylbiphenyl-3-carboxylic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4923-1G CA-4923-5G
COBBCA-4923-1GEA 0 GBP
COBBCA-4923-1G COBBCA-4923-5G
5'-Methoxy-2'-methylbiphenyl-3-carboxylic acid
5'-Methoxy-2'-methylbiphenyl-3-carboxylic acid
Formula: C₁₅H₁₄O₃
MDL Number: MFCD15143517
CAS Number: 1215205-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR