3'-Methoxy-4'-methylbiphenyl-3-carboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4921-5G
CA-4921-1G
COBBCA-4921-5GEA
0
GBP
COBBCA-4921-5G
COBBCA-4921-1G
3'-Methoxy-4'-methylbiphenyl-3-carboxylic acid
3'-Methoxy-4'-methylbiphenyl-3-carboxylic acid
Formula:
C₁₅H₁₄O₃ |
MDL Number:
MFCD15143515 CAS Number: 1215206-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...