2'-Methoxybiphenyl-3-carboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4819-5G
CA-4819-1G
COBBCA-4819-5GEA
0
GBP
COBBCA-4819-5G
COBBCA-4819-1G
2'-Methoxybiphenyl-3-carboxylic acid
2'-Methoxybiphenyl-3-carboxylic acid
Formula:
C₁₄H₁₂O₃ |
MDL Number:
MFCD03424597 CAS Number: 168618-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...