2'-Methoxy-5'-methylbiphenyl-3-carboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4914-1G
CA-4914-250M
COBBCA-4914-1GEA
0
GBP
COBBCA-4914-1G
COBBCA-4914-250M
2'-Methoxy-5'-methylbiphenyl-3-carboxylic acid
2'-Methoxy-5'-methylbiphenyl-3-carboxylic acid
Formula:
C₁₅H₁₄O₃ |
MDL Number:
MFCD11895636 CAS Number: 1181381-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...